Assay Details
Gene Symbol: CIPK04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ901232.1 | - | 4 - 5 | 344 | 75 | |
FR669159.1 | - | 3 - 4 | 317 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
