Assay Details
| Gene Symbol: CIPK018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ901246.1 | - | 1 - 2 | 170 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CIPK018| Gene Symbol: CIPK018 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FJ901246.1 | - | 1 - 2 | 170 | 95 | |
Set Membership: |
|