Assay Details
| Gene Symbol: PDS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | GQ221024.1 | - | 2 - 3 | 296 | 90 | |
| GQ221025.1 | - | 2 - 3 | 296 | 90 | ||
| GQ221026.1 | - | 2 - 3 | 296 | 90 | ||
| GQ221027.1 | - | 2 - 3 | 296 | 90 | ||
More Information
Set Membership: |
|

PDS1
Genomic Map