Assay Details
Gene Symbol: mindy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001204.1 | NP_001001204.1 | 8 - 9 | 890 | 66 | |
XM_018094601.1 | XP_017950090.1 | 8 - 9 | 890 | 66 | ||
XM_018094602.1 | XP_017950091.1 | 7 - 8 | 726 | 66 | ||
GenBank mRNA | BC067311.1 | - | 8 - 9 | 890 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
