Assay Details
| Gene Symbol: rhob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001214.1 | NP_001001214.1 | 1 | 555 | 67 | |
| GenBank mRNA | BC067992.1 | - | 1 | 555 | 67 | |
| CT025349.2 | - | 1 | 586 | 67 | ||
More Information
Set Membership: |
|

rhob
Genomic Map