Assay Details
Gene Symbol: hspa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001229.2 | NP_001001229.1 | 3 - 4 | 319 | 62 | |
GenBank mRNA | BC067910.2 | - | 3 - 4 | 319 | 62 | |
CR760959.2 | - | 3 - 4 | 256 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
