Assay Details
| Gene Symbol: srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001248.1 | NP_001001248.1 | 2 - 3 | 287 | 66 | |
| GenBank mRNA | BC068213.1 | - | 2 - 3 | 287 | 66 | |
More Information
Set Membership: |
|

srsf6
Genomic Map