Assay Details
| Gene Symbol: rtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001254.1 | NP_001001254.1 | 6 - 7 | 600 | 66 | |
| XM_012971620.2 | XP_012827074.1 | 6 - 7 | 683 | 66 | ||
| GenBank mRNA | BC067927.1 | - | 6 - 7 | 600 | 66 | |
More Information
Set Membership: |
|

rtf2
Genomic Map