Assay Details
| Gene Symbol: rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001857.1 | NP_001001857.1 | 4 - 5 | 543 | 58 | |
| XM_018097693.1 | XP_017953182.1 | 5 - 6 | 705 | 58 | ||
| GenBank mRNA | AY612091.1 | - | 4 - 5 | 543 | 58 | |
| BC136243.1 | - | 4 - 5 | 512 | 58 | ||
More Information
Set Membership: |
|

rnf114
Genomic Map