Assay Details
Gene Symbol: slc25a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004781.1 | NP_001004781.1 | 8 - 9 | 903 | 66 | |
GenBank mRNA | BC074516.1 | - | 8 - 9 | 903 | 66 | |
CR926214.2 | - | 8 - 9 | 807 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
