Assay Details
Gene Symbol: srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004783.1 | NP_001004783.1 | 4 - 5 | 400 | 82 | |
XM_012968191.2 | XP_012823645.1 | 5 - 6 | 498 | 82 | ||
GenBank mRNA | BC074518.1 | - | 4 - 5 | 400 | 82 | |
CR942775.1 | - | 4 - 5 | 401 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
