Assay Details
| Gene Symbol: stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004792.1 | NP_001004792.1 | 4 - 5 | 535 | 66 | |
| XM_012970284.2 | XP_012825738.1 | 4 - 5 | 933 | 66 | ||
| XM_018097075.1 | XP_017952564.1 | 4 - 5 | 595 | 66 | ||
| GenBank mRNA | BC074528.1 | - | 4 - 5 | 535 | 66 | |
More Information
Set Membership: |
|

stk16
Genomic Map