Assay Details
| Gene Symbol: cdc34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004793.1 | NP_001004793.1 | 1 - 2 | 341 | 107 | |
| GenBank mRNA | BC074529.1 | - | 1 - 2 | 341 | 107 | |
| CR760992.2 | - | 1 - 2 | 204 | 107 | ||
More Information
Set Membership: |
|

cdc34
Genomic Map