Assay Details
Gene Symbol: ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004800.1 | NP_001004800.1 | 200 | 104 | ||
GenBank mRNA | BC074550.1 | - | 200 | 104 | ||
CR760927.2 | - | 159 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
