Assay Details
| Gene Symbol: aimp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004804.1 | NP_001004804.1 | 3 - 4 | 580 | 75 | |
| XM_018097076.1 | XP_017952565.1 | 3 - 4 | 566 | 75 | ||
| GenBank mRNA | BC074561.1 | - | 3 - 4 | 580 | 75 | |
| CR760281.2 | - | 3 - 4 | 569 | 75 | ||
More Information
Set Membership: |
|

aimp2
Genomic Map