Assay Details
Gene Symbol: gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004805.1 | NP_001004805.1 | 1 - 2 | 154 | 68 | |
GenBank mRNA | BC074563.1 | - | 1 - 2 | 154 | 68 | |
CR855824.1 | - | 1 - 2 | 140 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
