Assay Details
| Gene Symbol: eif1ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004814.1 | NP_001004814.1 | 2 - 3 | 340 | 70 | |
| GenBank mRNA | BC074588.1 | - | 2 - 3 | 340 | 70 | |
| CR760442.2 | - | 2 - 3 | 140 | 70 | ||
More Information
Set Membership: |
|

eif1ax
Genomic Map