Assay Details
Gene Symbol: eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004814.1 | NP_001004814.1 | 4 - 5 | 496 | 87 | |
GenBank mRNA | BC074588.1 | - | 4 - 5 | 496 | 87 | |
CR760442.2 | - | 4 - 5 | 296 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
