Assay Details
Gene Symbol: tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004819.1 | NP_001004819.1 | 4 - 5 | 663 | 74 | |
XM_012964508.2 | XP_012819962.1 | 4 - 5 | 702 | 74 | ||
XM_012964509.2 | XP_012819963.1 | 4 - 5 | 702 | 74 | ||
XM_012964511.2 | XP_012819965.1 | 4 - 5 | 702 | 74 | ||
XM_012964512.2 | XP_012819966.1 | 5 - 6 | 1011 | 74 | ||
XM_018094613.1 | XP_017950102.1 | 4 - 5 | 1258 | 74 | ||
GenBank mRNA | BC074599.1 | - | 4 - 5 | 663 | 74 | |
CR761302.2 | - | 4 - 5 | 594 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
