Assay Details
| Gene Symbol: msantd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004822.1 | NP_001004822.1 | 2 - 3 | 706 | 94 | |
| XM_012956711.2 | XP_012812165.1 | 2 - 3 | 3371 | 94 | ||
| GenBank mRNA | BC074602.1 | - | 2 - 3 | 706 | 94 | |
| CR855752.2 | - | 2 - 3 | 638 | 94 | ||
More Information
Set Membership: |
|

msantd4
Genomic Map