Assay Details
| Gene Symbol: rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004840.1 | NP_001004840.1 | 3 - 4 | 394 | 74 | |
| GenBank mRNA | BC074649.1 | - | 3 - 4 | 394 | 74 | |
| CR760977.2 | - | 3 - 4 | 343 | 74 | ||
More Information
Set Membership: |
|

rac1
Genomic Map