Assay Details
| Gene Symbol: nr2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004841.1 | NP_001004841.1 | 2 - 3 | 362 | 76 | |
| XM_012959039.2 | XP_012814493.1 | 2 - 3 | 545 | 76 | ||
| XM_018091958.1 | XP_017947447.1 | 2 - 3 | 486 | 76 | ||
| GenBank mRNA | BC074651.1 | - | 2 - 3 | 362 | 76 | |
More Information
Set Membership: |
|

nr2f6
Genomic Map