Assay Details
Gene Symbol: imp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004848.1 | NP_001004848.1 | 3 - 4 | 479 | 80 | |
XM_018092989.1 | XP_017948478.1 | 3 - 4 | 261 | 80 | ||
GenBank mRNA | BC074666.1 | - | 3 - 4 | 479 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
