Assay Details
Gene Symbol: nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004851.1 | NP_001004851.1 | 1 - 2 | 354 | 80 | |
GenBank mRNA | BC074672.1 | - | 1 - 2 | 354 | 80 | |
CR760347.2 | - | 1 - 2 | 343 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
