Assay Details
| Gene Symbol: eif5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004855.1 | NP_001004855.1 | 2 - 3 | 295 | 92 | |
| XM_012962865.1 | XP_012818319.1 | 3 - 4 | 465 | 92 | ||
| XM_012962866.2 | XP_012818320.1 | 3 - 4 | 396 | 92 | ||
| XM_012962867.1 | XP_012818321.1 | 3 - 4 | 321 | 92 | ||
| XM_012962868.2 | XP_012818322.1 | 3 - 4 | 314 | 92 | ||
| XM_012962869.2 | XP_012818323.1 | 3 - 4 | 310 | 92 | ||
| GenBank mRNA | BC074676.1 | - | 2 - 3 | 295 | 92 | |
| CR760910.2 | - | 2 - 3 | 275 | 92 | ||
More Information
Set Membership: |
|

eif5a
Genomic Map