Assay Details
| Gene Symbol: nudt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004857.1 | NP_001004857.1 | 2 - 3 | 272 | 101 | |
| XM_012970291.2 | XP_012825745.1 | 2 - 3 | 101 | |||
| XM_012970292.2 | XP_012825746.1 | 2 - 3 | 101 | |||
| GenBank mRNA | BC074680.1 | - | 2 - 3 | 272 | 101 | |
More Information
Set Membership: |
|

nudt1
Genomic Map