Assay Details
Gene Symbol: chp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004859.1 | NP_001004859.1 | 2 - 3 | 174 | 80 | |
XM_012968198.2 | XP_012823652.1 | 2 - 3 | 239 | 80 | ||
GenBank mRNA | BC074687.1 | - | 2 - 3 | 174 | 80 | |
CR855591.2 | - | 2 - 3 | 152 | 80 | ||
CR926234.1 | - | 2 - 3 | 156 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
