Assay Details
| Gene Symbol: nectin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004860.1 | NP_001004860.1 | 4 - 5 | 938 | 101 | |
| XM_012956717.2 | XP_012812171.1 | 4 - 5 | 987 | 101 | ||
| GenBank mRNA | BC074691.1 | - | 4 - 5 | 938 | 101 | |
| CR762185.2 | - | 4 - 5 | 937 | 101 | ||
| Gene Symbol: LOC108645444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018090656.1 | XP_017946145.1 | 3 - 4 | 757 | 101 | |
More Information
Set Membership: |
|

nectin2
Genomic Map