Assay Details
| Gene Symbol: chmp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004866.1 | NP_001004866.1 | 2 - 3 | 459 | 80 | |
| GenBank mRNA | BC074708.1 | - | 2 - 3 | 459 | 80 | |
| CR762287.2 | - | 2 - 3 | 426 | 80 | ||
More Information
Set Membership: |
|

chmp4b
Genomic Map