Assay Details
| Gene Symbol: gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278521.1 | NP_001265450.1 | 5 - 6 | 545 | 101 | |
| XM_012968199.2 | XP_012823653.1 | 6 - 7 | 633 | 101 | ||
| GenBank mRNA | BC075259.1 | - | 5 - 6 | 467 | 101 | |
| CR760021.2 | - | 5 - 6 | 467 | 101 | ||
More Information
Set Membership: |
|

gnpnat1
Genomic Map