Assay Details
Gene Symbol: rab11b.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004880.1 | NP_001004880.1 | 3 - 4 | 439 | 90 | |
GenBank mRNA | BC075268.1 | - | 3 - 4 | 439 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
