Assay Details
| Gene Symbol: hmgb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004888.1 | NP_001004888.1 | 2 - 3 | 230 | 81 | |
| XM_018096198.1 | XP_017951687.1 | 2 - 3 | 267 | 81 | ||
| GenBank mRNA | BC075290.1 | - | 2 - 3 | 230 | 81 | |
| CR761259.2 | - | 2 - 3 | 259 | 81 | ||
More Information
Set Membership: |
|

hmgb3
Genomic Map