Assay Details
| Gene Symbol: arpin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004898.1 | NP_001004898.1 | 3 - 4 | 330 | 88 | |
| XM_012958820.2 | XP_012814274.1 | 4 - 5 | 483 | 88 | ||
| XM_012958821.2 | XP_012814275.1 | 4 - 5 | 472 | 88 | ||
| XM_012958822.2 | XP_012814276.1 | 3 - 4 | 301 | 88 | ||
| XM_012958823.2 | XP_012814277.1 | 3 - 4 | 293 | 88 | ||
| GenBank mRNA | BC075312.1 | - | 3 - 4 | 330 | 88 | |
More Information
Set Membership: |
|

arpin
Genomic Map