Assay Details
| Gene Symbol: hbg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004899.1 | NP_001004899.1 | 1 - 2 | 143 | 95 | |
| GenBank mRNA | BC075313.1 | - | 1 - 2 | 143 | 95 | |
More Information
Set Membership: |
|

hbg2
Genomic Map