Assay Details
Gene Symbol: nudt19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004903.1 | NP_001004903.1 | 1 - 2 | 808 | 100 | |
XM_012960826.2 | XP_012816280.1 | 3 - 4 | 1135 | 100 | ||
XM_012960828.2 | XP_012816282.1 | 3 - 4 | 1283 | 100 | ||
XM_012960829.2 | XP_012816283.1 | 3 - 4 | 1088 | 100 | ||
XM_012960830.1 | XP_012816284.1 | 2 - 3 | 862 | 100 | ||
XM_012960831.2 | XP_012816285.1 | 2 - 3 | 848 | 100 | ||
XM_012960832.2 | XP_012816286.1 | 2 - 3 | 848 | 100 | ||
XM_018092992.1 | XP_017948481.1 | 3 - 4 | 1083 | 100 | ||
GenBank mRNA | BC075322.1 | - | 1 - 2 | 808 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
