Assay Details
| Gene Symbol: mpv17l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004930.1 | NP_001004930.1 | 5 - 6 | 673 | 85 | |
| XM_012959523.2 | XP_012814977.1 | 4 - 5 | 445 | 85 | ||
| GenBank mRNA | BC075397.1 | - | 5 - 6 | 673 | 85 | |
| CR761343.2 | - | 5 - 6 | 664 | 85 | ||
More Information
Set Membership: |
|

mpv17l2
Genomic Map