Assay Details
Gene Symbol: tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004932.1 | NP_001004932.1 | 3 - 4 | 391 | 74 | |
GenBank mRNA | BC075401.1 | - | 3 - 4 | 391 | 74 | |
CR760634.2 | - | 2 - 3 | 262 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
