Assay Details
Gene Symbol: prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004941.1 | NP_001004941.1 | 2 - 3 | 240 | 78 | |
GenBank mRNA | BC075423.1 | - | 2 - 3 | 240 | 78 |
Gene Symbol: LOC100489438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002941034.2 | XP_002941080.1 | 2 - 3 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
