Assay Details
| Gene Symbol: prss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004941.1 | NP_001004941.1 | 3 - 4 | 499 | 76 | |
| GenBank mRNA | BC075423.1 | - | 3 - 4 | 499 | 76 | |
| Gene Symbol: LOC100489438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002941034.2 | XP_002941080.1 | 3 - 4 | 505 | 76 | |
More Information
Set Membership: |
|

prss3
Genomic Map