Assay Details
Gene Symbol: tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004962.1 | NP_001004962.1 | 2 - 3 | 750 | 97 | |
XM_012959738.2 | XP_012815192.1 | 2 - 3 | 1138 | 97 | ||
XM_012959746.2 | XP_012815200.1 | 2 - 3 | 743 | 97 | ||
XM_012959747.2 | XP_012815201.1 | 2 - 3 | 1065 | 97 | ||
XM_012959751.2 | XP_012815205.1 | 2 - 3 | 1161 | 97 | ||
GenBank mRNA | BC075471.1 | - | 2 - 3 | 750 | 97 | |
CT025209.2 | - | 1 - 2 | 703 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
