Assay Details
Gene Symbol: tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004962.1 | NP_001004962.1 | 3 - 4 | 879 | 87 | |
GenBank mRNA | BC075471.1 | - | 3 - 4 | 879 | 87 | |
CT025209.2 | - | 2 - 3 | 832 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
