Assay Details
Gene Symbol: sdr16c5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004963.1 | NP_001004963.1 | 2 - 3 | 490 | 72 | |
XM_012964524.2 | XP_012819978.1 | 2 - 3 | 509 | 72 | ||
GenBank mRNA | BC075474.1 | - | 2 - 3 | 490 | 72 | |
CT025330.2 | - | 2 - 3 | 467 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
