Assay Details
Gene Symbol: gstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004964.1 | NP_001004964.1 | 6 - 7 | 508 | 102 | |
GenBank mRNA | BC075475.1 | - | 6 - 7 | 508 | 102 | |
CR761400.2 | - | 6 - 7 | 528 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
