Assay Details
Gene Symbol: rpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004965.2 | NP_001004965.1 | 2 - 3 | 99 | 64 | |
XM_012954243.1 | XP_012809697.1 | 3 - 4 | 138 | 64 | ||
GenBank mRNA | BC075477.1 | - | 2 - 3 | 103 | 64 | |
BC158397.1 | - | 2 - 3 | 113 | 64 | ||
CU025127.1 | - | 2 - 3 | 98 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
