Assay Details
| Gene Symbol: abitram | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004966.1 | NP_001004966.1 | 1 - 2 | 129 | 100 | |
| XM_012964525.2 | XP_012819979.1 | 2 - 3 | 264 | 100 | ||
| XM_012964526.2 | XP_012819980.1 | 2 - 3 | 257 | 100 | ||
| XM_012964527.2 | XP_012819981.1 | 2 - 3 | 254 | 100 | ||
| GenBank mRNA | BC075478.1 | - | 1 - 2 | 129 | 100 | |
| CR761531.2 | - | 2 - 3 | 169 | 100 | ||
More Information
Set Membership: |
|

abitram
Genomic Map