Assay Details
| Gene Symbol: abitram | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004966.1 | NP_001004966.1 | 4 - 5 | 468 | 96 | |
| XM_012964525.2 | XP_012819979.1 | 5 - 6 | 603 | 96 | ||
| GenBank mRNA | BC075478.1 | - | 4 - 5 | 468 | 96 | |
More Information
Set Membership: |
|

abitram
Genomic Map