Assay Details
Gene Symbol: abitram | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004966.1 | NP_001004966.1 | 5 - 6 | 550 | 116 | |
XM_012964525.2 | XP_012819979.1 | 6 - 7 | 685 | 116 | ||
XM_012964526.2 | XP_012819980.1 | 5 - 6 | 519 | 116 | ||
XM_012964527.2 | XP_012819981.1 | 5 - 6 | 516 | 116 | ||
GenBank mRNA | BC075478.1 | - | 5 - 6 | 550 | 116 | |
CR761531.2 | - | 5 - 6 | 431 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
