Assay Details
Gene Symbol: ankrd40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004991.1 | NP_001004991.1 | 3 - 4 | 384 | 73 | |
XM_012971643.2 | XP_012827097.1 | 2 - 3 | 729 | 73 | ||
XM_012971644.2 | XP_012827098.1 | 3 - 4 | 532 | 73 | ||
XM_012971645.2 | XP_012827099.1 | 3 - 4 | 521 | 73 | ||
XM_012971646.2 | XP_012827100.1 | 3 - 4 | 500 | 73 | ||
GenBank mRNA | BC075561.1 | - | 3 - 4 | 384 | 73 | |
CR761248.1 | - | 2 - 3 | 536 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
