Assay Details
| Gene Symbol: gtf2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005005.1 | NP_001005005.1 | 4 - 5 | 913 | 94 | |
| GenBank mRNA | BC076651.1 | - | 4 - 5 | 913 | 94 | |
More Information
Set Membership: |
|

gtf2e1
Genomic Map