Assay Details
| Gene Symbol: ncbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005015.1 | NP_001005015.1 | 8 - 9 | 992 | 66 | |
| GenBank mRNA | BC076682.1 | - | 8 - 9 | 992 | 66 | |
More Information
Set Membership: |
|

ncbp3
Genomic Map